Order Entry
Czech Republic
ContactUsLinkComponent
Phenyl trifluoroacetate 98%
Flammable
Irritant
Phenyl trifluoroacetate 98%
Catalog # APOSPC6223-100G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
Phenyl trifluoroacetate 98%
Catalog # APOSPC6223-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :190.12 g/mol
Boiling Pt:146 - 147 °C
Boiling Pt:146…147 °C
Density:1.276
Flash Pt:52 °C
UN:3272
ADR:3,III
 :GHS02
 :GHS07
CAS Number:500-73-2
Formula:C₈H₅F₃O₂
MDL Number:MFCD00039232