
Methyl perfluoro(2-methyl-3-oxahexanoate) 97%
Catalog # APOSPC6122D-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:344.079 g/mol
Boiling Pt:115 - 117 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:13140-34-6
Formula:C₇H₃F₁₁O₃
MDL Number:MFCD00042358
Boiling Pt:115…117 °C