
2-Bromo-5-fluoro-3-(trifluoromethyl)benzeneboronic acid 95%
:
:C₇H₄BBrF₄O₂
:MFCD32903692
:286.81 g/mol
:GHS07
Frequently Bought Together
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Water, TECHNICAL, demineralised
: VWR Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals