
2-Bromo-5-(difluoromethyl)benzoic acid 95%
:1783329-70-3
:
:C₈H₅BrF₂O₂
:MFCD32070685
:251.02 g/mol
:GHS07
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
1-Chloro-2,4-dinitrobenzene 99%
: Thermo Fisher Scientific
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 98%, pure
: Thermo Fisher Scientific
n-Heptane, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
N,N-DIMETHYLFORMAMIDE AR | 2,5 DM3 1 * 2,5 L
: MACRON AVANTOR BRAND
Methyl ethyl ketone
: Lach-ner S.R.O.
Cyclohexane 99.8% for HPLC
: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
: Thermo Fisher Scientific