Order Entry
Czech Republic
ContactUsLinkComponent
4-Chloro-5-fluoropyridine-3-carboxylic Acid 95%
4-Chloro-5-fluoropyridine-3-carboxylic Acid 95%
Catalog # APOSPC57470-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-5-fluoropyridine-3-carboxylic Acid 95%
Catalog # APOSPC57470-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.545 g/mol
CAS Number:860296-20-4
Formula:C₆H₃ClFNO₂
MDL Number:MFCD18257385