
5-Chloro-4-fluoro-2-(trifluoromethyl)benzyl bromide 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2090498-26-1
:
Formula:C₈H₄BrClF₄
MDL Number:MFCD31696997
:291.47 g/mol
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together