
5-(Difluoromethyl)pyridin-3-amine, 3-BOC protected 95%
Catalog # APOSPC55856-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:244.24 g/mol
:GHS07
CAS Number:899899-70-8
Formula:C₁₁H₁₄F₂N₂O₂
MDL Number:MFCD27981250