
5-Nitro-2-(tetrahydro-2H-pyran-4-ylmethoxy)pyridine
Catalog # APOSOR111004-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:238.25 g/mol
:GHS07
CAS Number:1287218-29-4
Formula:C₁₁H₁₄N₂O₄
MDL Number:MFCD14666511