
3-(Dimethylamino)-1-(2,4,5-trichlorophenyl)prop-2-en-1-one
Catalog # APOSOR70231-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:278.56 g/mol
:GHS07
CAS Number:2645406-32-0
Formula:C₁₁H₁₀Cl₃NO
MDL Number:MFCD32862647