
Triphenylphosphine polymer bound 1.4 - 2.0 mmol/g on polystyrene, cross-linked with 1% DVB 200 - 400 mesh
Supplier: Apollo Scientific
Certificates

Triphenylphosphine polymer bound 1.4 - 2.0 mmol/g on polystyrene, cross-linked with 1% DVB 200 - 400 mesh
Supplier: Apollo Scientific
About this item
CAS Number:39319-11-4
:
Formula:C₁₈H₁₅P
MDL Number:MFCD20489348
:262.292 g/mol
:GHS07
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals