
3-Bromo-2-chlorobenzoic acid
:56961-27-4
:
:C₇H₄BrClO₂
:MFCD01569399
:163…170 °C
:235.46 g/mol
:GHS07
Frequently Bought Together
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
: Thermo Fisher Scientific