Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-DL-pyroglutamate 95%
Irritant
Methyl-DL-pyroglutamate 95%
Catalog # APOSOR928083-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-DL-pyroglutamate 95%
Catalog # APOSOR928083-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :143.14 g/mol
Boiling Pt:133 - 135/1 mm °C
Boiling Pt:133…135/1 mm °C
Melting Pt:21 - 23 °C
Melting Pt:21…23 °C
Density:1.25
 :GHS07
CAS Number:54571-66-3
Formula:C₆H₉NO₃
MDL Number:MFCD03844651