


4-Chloro-3-iodobenzotrifluoride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:672-57-1
:
Formula:C₇H₃ClF₃I
MDL Number:MFCD00001037
:306.45 g/mol
Boiling Pt:58…60/0.5 mm °C
Density:1.973
Flash Pt:>110 °C
UN:3265
ADR:8,II
:GHS05
:GHS09
:GHS06
Frequently Bought Together