
N-Methyl-N-[(methylamino)thioxomethyl]carbamic acid, phenyl ester 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1562489-90-0
:
Formula:C₁₀H₁₂N₂O₂S
MDL Number:MFCD34561781
:224.28 g/mol
:GHS07
Frequently Bought Together