
2-(Trifluoromethyl)-1H-benzimidazol-4-amine
Catalog # APOSPC32933-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:201.15 g/mol
UN:2811
:GHS07
CAS Number:189045-18-9
Formula:C₈H₆F₃N₃
MDL Number:MFCD05856872