4-Fluoro-6-(trifluoromethyl)-1H-benzimidazole
Catalog # APOSPC500991-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:204.12 g/mol
CAS Number:1360891-34-4
Formula:C₈H₄F₄N₂
MDL Number:MFCD28154923