

Bis(diphenylphosphino)methane 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2071-20-7
:
Formula:C₂₅H₂₂P₂
MDL Number:MFCD00003537
Melting Pt:118…122 °C
:384.4 g/mol
:GHS07
:GHS09
Frequently Bought Together
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Ethyl-1-hexanol 99%
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, extra pure
Supplier: Thermo Fisher Scientific
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Kerosene, Purum
Supplier: Honeywell Chemicals
Dibutylamine for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Diethyl ether stabilised, BAKER, J.T.Baker®
Supplier: Avantor