
5-Chloro-2-nitrotoluene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5367-28-2
:
Formula:C₇H₆ClNO₂
MDL Number:MFCD00024579
:171.58 g/mol
Melting Pt:27…30 °C
UN:3457
ADR:6.1,III
:GHS07
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Dichloromethane, BAKER ANALYZED® HPLC for HPLC, J.T.Baker®
Supplier: Avantor
Acetonitrile, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahexylammonium bromide, Supelco®
Supplier: Merck
Acetone, J.T.Baker®
Supplier: Avantor
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene ≥99.8% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Formamide ≥99.5% (by GC, titration analysis), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Carbon disulphide 99.9% ACS
Supplier: Thermo Fisher Scientific