Order Entry
Czech Republic
ContactUsLinkComponent
3-(2-Chlorophenyl)-1-propene
3-(2-Chlorophenyl)-1-propene
Catalog # APOSOR01876-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-(2-Chlorophenyl)-1-propene
Catalog # APOSOR01876-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1-Chloro-2-(2-propen-1-yl)benzene, 1-Allyl-2-chlorobenzene, 1-Chloro-2-(prop-2-en-1-yl)benzene, 3-(2-Chlorophenyl)prop-1-ene
 :
 :152.63 g/mol
 :GHS07
CAS Number:1587-07-1
Formula:C₉H₉Cl
MDL Number:MFCD00135025