Pesticide reference standard, 2,3,4,5-Tetrachloroanisole 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL17333100IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:938-86-3
- Description:2,3,4,5-Tetrachloroanisole 10 µg/ml in isooctane
- Formula:C₇H₄Cl₄O
- Description:2,3,4,5-Tetrachloroanisole