Order Entry
Czech Republic
ContactUsLinkComponent
4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionic Acid [Bodi Fluor™ 488 acid] ≥95%
Irritant
4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionic Acid [Bodi Fluor™ 488 acid] ≥95%
Catalog # AATB700
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionic Acid [Bodi Fluor™ 488 acid] ≥95%
Catalog # AATB700
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :292.09 g/mol
ADR:5,II
 :GHS07
CAS Number:126250-45-1
Formula:C₁₄H₁₅BF₂N₂O₂