Order Entry
Czech Republic
ContactUsLinkComponent
4-Methylumbelliferone fluorescence reference standard
Irritant
4-Methylumbelliferone fluorescence reference standard
Catalog # AATB41
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
4-Methylumbelliferone fluorescence reference standard
Catalog # AATB41
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :176.17 g/mol
ADR:5,II
 :GHS07
CAS Number:90-33-5
Formula:C₁₀H₈O₃
MDL Number:MFCD00006866