Order Entry
Czech Republic
ContactUsLinkComponent
5-Carboxy-4,7-dichloror-X-hodamine (5-dROX) ≥95%
5-Carboxy-4,7-dichloror-X-hodamine (5-dROX) ≥95%
  AATB310
 :  
undefined
5-Carboxy-4,7-dichloror-X-hodamine (5-dROX) ≥95%
  AATB310
 :  

 

  • Pk:
    25 mg
  • Seq:
    2315

 

 

 :
 :704.69 g/mol