Order Entry
Czech Republic
ContactUsLinkComponent
 
Cyanine 3.5 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy3.5® NHS ester]
Catalog # AATB275
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cyanine 3.5 monosuccinimidyl ester potassium salt ≥90% [same as GE Cy3.5® NHS ester]
Catalog # AATB275
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1102.36 g/mol