5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF) mixture of isomers ≥95%
Catalog # AATB235
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:445.204 g/mol
CAS Number:111843-78-8
Formula:C₂₁H₁₀Cl₂O₇
MDL Number:MFCD00037452