0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Tide Fluor™ 4 alkyne
Tide Fluor™ 4 alkyne, Tide Fluor™ fluorescent probe
Tide Fluor™ 4 alkyne, Tide Fluor™ fluorescent probe
Irritant
Tide Fluor™ 4 alkyne, Tide Fluor™ fluorescent probe
Catalog # AATB2301
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
Tide Fluor™ 4 alkyne, Tide Fluor™ fluorescent probe
Catalog # AATB2301
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
599.72 g/mol
ADR:
5,II
:
GHS07
Product Details