5(6)-Carboxyfluorescein diacetate mixture of isomers ≥95%
Catalog # AATB22021
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:460.39 g/mol
CAS Number:124387-19-5
Formula:C₂₅H₁₆O₉
MDL Number:MFCD00467494