
6-Methoxy-N-(3-sulphopropyl)quinolinium (SPQ) chloride indicator
Catalog # AATB21252
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:281 g/mol
ADR:5,II
:GHS07
CAS Number:83907-40-8
Formula:C₁₅H₁₅NO₄S
MDL Number:MFCD00037592