Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
FLUO-3 pentasodium salt
FLUO-3 pentasodium salt
FLUO-3 pentasodium salt
AATB21016
:
AAT BIOQUEST
:
FLUO-3 pentasodium salt
AATB21016
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
5,II
:
879 g/mol
:
GHS07
:
134907-84-9
:
C₃₆H₂₅Cl₂Na₅N₂O₁₃