
Tide Quencher™ 5WS succinimidyl ester, Tide Quencher™ fluorescent probe
Catalog # AATB2081
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:974.24 g/mol
ADR:5,II
:GHS07