Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ICG-Sulpho-OSu
ICG-Sulpho-OSu ≥80%
ICG-Sulpho-OSu ≥80%
ICG-Sulpho-OSu ≥80%
AATB180
:
AAT BIOQUEST
:
ICG-Sulpho-OSu ≥80%
AATB180
:
AAT BIOQUEST
:
Pk:
1 mg
Seq:
2070
Environmentally Preferable:
:
:
190714-28-4