Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aminopropargyl dATP
Aminopropargyl dATP for PCR
Aminopropargyl dATP for PCR
Irritant
Aminopropargyl dATP for PCR
Catalog # AATB17056
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
Aminopropargyl dATP for PCR
Catalog # AATB17056
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
10 µmol
Seq:
9105
Specifications
About this item
Product Details
:
:
566.99 g/mol
ADR:
5,II
:
GHS07
CAS Number:
587848-72-4
Product Details