Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Biotin-11-dGTP
Biotin-11-dGTP 1 mM in TRIS buffer (pH 7.5)
Biotin-11-dGTP 1 mM in TRIS buffer (pH 7.5)
Irritant
Biotin-11-dGTP 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17015
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
Biotin-11-dGTP 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17015
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Typ balení:
Plastic bottle
Bal.:
25 Nmo
Sekvence:
0311
Specifications
About this item
Detaily produktu
:
:
898.71 g/mol
ADR:
5,II
:
GHS07
Detaily produktu