Order Entry
Czech Republic
ContactUsLinkComponent
Digoxigenin-11-dUTP 1 mM for PCR
Irritant
Digoxigenin-11-dUTP 1 mM for PCR
Catalog # AATB17012
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Digoxigenin-11-dUTP 1 mM for PCR
Catalog # AATB17012
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 Nmo
  • Seq:
    0311

Specifications

About this item

 :
 :1132.87 g/mol
ADR:5,II
 :GHS07
CAS Number:129030-36-0