Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
TF5-dUTP 1 mM in TRIS buffer (pH 7.5)
Katalogové číslo AATB17010
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
TF5-dUTP 1 mM in TRIS buffer (pH 7.5)
Katalogové číslo AATB17010
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 Nmo
  • Seq:
    0311

Specifications

About this item

 :
 :1570 g/mol
ADR:5,II
 :GHS07