0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
MeO-Succ-Arg-Pro-Tyr-AMC
MeO-Succ-Arg-Pro-Tyr-AMC
MeO-Succ-Arg-Pro-Tyr-AMC
Irritant
MeO-Succ-Arg-Pro-Tyr-AMC
Catalog # AATB13460
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
MeO-Succ-Arg-Pro-Tyr-AMC
Catalog # AATB13460
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details