Order Entry
Czech Republic
ContactUsLinkComponent
NTA maleimide for thiol-surface biomolecular attachment
Irritant
NTA maleimide for thiol-surface biomolecular attachment
Catalog # AATB12609
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
NTA maleimide for thiol-surface biomolecular attachment
Catalog # AATB12609
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :413.38 g/mol
ADR:5,II
 :GHS07
CAS Number:935666-21-0