Anti-IgG Goat Polyclonal Antibody [clone: IgG] (FITC (Fluorescein Isothiocyanate))
Catalog # AATB16876
Supplier: AAT BIOQUEST
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Clone:IgG
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Goat
- Isotype:IgG (H, L)
- Reactivity:Rabbit
- Environmentally Preferable:
- Cross adsorption:Yes
- Pack type:Vial
- Pk:1 mg
Specifications
About this item
AAT Bioquest's anti-mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target.
- Affinity-purified
- Bright signal
- Cross-adsorbed
Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse