ContactUsLinkComponent
TRITC-Dextran Conjugate (Average MW = ~70K)
TRITC-Dextran Conjugate (Average MW = ~70K)
Catalog # AATB21708
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
TRITC-Dextran Conjugate (Average MW = ~70K)
Catalog # AATB21708
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    25 mg
  • Description:
    TRITC–dextran conjugate (average MW = ~70K)
Specifications
About this item
TRITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.
  • High quality
  • Multiple applications
Caution: For research use only.