ContactUsLinkComponent
Tetracysteine FLASH Diacetate *2 mM DMSO Solution*
Tetracysteine FLASH Diacetate *2 mM DMSO Solution*
Catalog # AATB22334
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Tetracysteine FLASH Diacetate *2 mM DMSO Solution*
Catalog # AATB22334
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Tetracysteine FLASH diacetate *2 mM DMSO solution*
  • Tests per kit:
    100 Tests
Specifications
About this item
FLASH diacetate is a cell-permeable version of FlASH, which is a fluorescein derivative, modified to contain two arsenic atoms at a set distance from each other.
  • Fluorescein derivative
  • High affinity for thiols
  • Multiple applications
Caution: For research use only.