ContactUsLinkComponent
ReadiUse™ Preactivated HRP NHS Ester
ReadiUse™ Preactivated HRP NHS Ester
Catalog # AATB11025
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiUse™ Preactivated HRP NHS Ester
Catalog # AATB11025
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    300 µG
  • Description:
    ReadiUse™ preactivated HRP NHS ester
  • Environmentally Preferable:
Specifications
About this item
ReadiUse™ Preactivated HRP NHS ester is the mono NHS ester of HRP.
  • Ready-to-use
  • Highly useful antibody label
  • Widely used for immunoassays
  • Multiple applications
Caution: For research use only.