Order Entry
Czech Republic
ContactUsLinkComponent
 
ReadiUse™ dNTP Mix *10 mM* (5 ml)
  AATB17200
 :  
undefined
ReadiUse™ dNTP Mix *10 mM* (5 ml)
  AATB17200
 :  

 

  • Pk:
    5 mL
  • Description:
    ReadiUse™ dNTP Mix *10 mm*

 

 

Our Ready-to Use Solution of deoxynucleotide triphosphates (dNTPs) have been extensively tested and verified for use in a wide variety of molecular biology applications, including PCR, real-time PCR, high-fidelity PCR, long PCR, cDNA synthesis, RT-PCR, RCA, MDA, DNA labelling, DNA sequencing, in vitro transcription, siRNA synthesis, and RNA amplification.

  • Ready to use formulation
  • In-house testing and quality control
  • Multiple applications
 : For research use only.