ContactUsLinkComponent
Amplite® Fluorimetric Glutathione GSH/GSSG Ratio Assay Kit *Green Fluorescence*
Amplite® Fluorimetric Glutathione GSH/GSSG Ratio Assay Kit *Green Fluorescence*
Catalog # AATB10056
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Fluorimetric Glutathione GSH/GSSG Ratio Assay Kit *Green Fluorescence*
Catalog # AATB10056
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ fluorimetric glutathione GSH/GSSG ratio assay kit *green fluorescence*
  • Tests per kit:
    200 Tests
Specifications
About this item
When cells are exposed to increased levels of oxidative stress, GSSG will accumulate and the ratio of GSH to GSSG will decrease.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.