Zadání objednávky
Czech Republic
ContactUsLinkComponent
Cell Meter™ Intracellular Fluorimetric Hydrogen Peroxide Assay Kit *Blue Fluorescence*
Cell Meter™ Intracellular Fluorimetric Hydrogen Peroxide Assay Kit *Blue Fluorescence*
Catalog # AATB11504
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ Intracellular Fluorimetric Hydrogen Peroxide Assay Kit *Blue Fluorescence*
Catalog # AATB11504
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Bal.:
    100 Tests
  • Popis:
    Cell Meter™ intracellular fluorimetric hydrogen peroxide assay kit *blue fluorescence*
Specifications
About this item
Hydrogen peroxide is a reactive oxygen metabolic by-product that serves as a key regulator for a number of oxidative stress-related states.
  • Robust protocol
  • High sensitivity
  • Quantifies H₂O₂ in live cells
Caution: For research use only.