ContactUsLinkComponent
Amplite® Colorimetric Glucose-6-Phosphate Dehydrogenase (G6PD) Assay Kit
Amplite® Colorimetric Glucose-6-Phosphate Dehydrogenase (G6PD) Assay Kit
Catalog # AATB13807
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Colorimetric Glucose-6-Phosphate Dehydrogenase (G6PD) Assay Kit
Catalog # AATB13807
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ colorimetric glucose-6-phosphate dehydrogenase (G6PD) assay kit
Specifications
About this item
Glucose-6-phosphate dehydrogenase (G6PD) catalyses the conversion of glucose-6-phosphate to 6-phosphoglucono-δ-lactone, the first and rate-limiting step in the pentose phosphate pathway.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.