ContactUsLinkComponent
Cell Meter™ Live Cell Caspase 3/7 Binding Assay Kit *Red Fluorescence*
Cell Meter™ Live Cell Caspase 3/7 Binding Assay Kit *Red Fluorescence*
Catalog # AATB20101
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ Live Cell Caspase 3/7 Binding Assay Kit *Red Fluorescence*
Catalog # AATB20101
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    25 Tests
  • Description:
    Cell Meter™ live cell caspase 3/7 binding assay kit *Red fluorescence*
Specifications
About this item
The Cell Meter™ live cell caspases activity assay kits are based on fluorescent FMK inhibitors of caspases.
  • Based on FMK inhibitor
  • Direct caspase detection
  • Suitable for all major fluorescence platforms
Caution: For research use only.