Cell Meter™ NIR Mitochondrion Membrane Potential Assay Kit *Optimized for Flow Cytometry*
Catalog # AATB22802
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:100 Tests
- Description:Cell Meter™ NIR mitochondrion membrane potential assay kit *optimized for flow cytometry*
Specifications
About this item
The Cell Meter™ assay kits are a set of tools for monitoring cell viability.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimised for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.