ContactUsLinkComponent
Amplite® Colorimetric α-Ketoglutarate Quantitation Kit
Amplite® Colorimetric α-Ketoglutarate Quantitation Kit
Catalog # AATB10085
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Colorimetric α-Ketoglutarate Quantitation Kit
Catalog # AATB10085
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ colorimetric α-ketoglutarate quantitation kit
Specifications
About this item
Alpha-ketoglutarate (α-Ketoglutarate) is a key molecule in the Krebs cycle determining the overall rate of the citric acid cycle of the organism.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
This colorimetric α-Ketoglutarate Quantitation Kit offers a sensitive colorimetric assay for quantifying α-ketoglutarate in biological samples.
Caution: For research use only.