Order Entry
Czech Republic
ContactUsLinkComponent
MycoLight™ Ratiometric Bacterial Membrane Potential Kit *Red/Green Fluorescence*
MycoLight™ Ratiometric Bacterial Membrane Potential Kit *Red/Green Fluorescence*
Catalog # AATB22401
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
MycoLight™ Ratiometric Bacterial Membrane Potential Kit *Red/Green Fluorescence*
Catalog # AATB22401
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    MycoLight™ ratiometric bacterial membrane potential kit *red/green fluorescence*
  • Environmentally Preferable:

Specifications

About this item

AAT Bioquest's MycoLight™ Ratiometric Bacterial Membrane Potential Kit uses a fluorescent sensor that exhibits green fluorescence in both gram-positive and negative bacterial cells in low concentration, but the fluorescence shifts toward red emission at higher cytosolic concentrations due to the dye molecule aggregation caused by larger membrane potentials.

  • Suitable for gram-positive and gram-negative bacteria
  • Ratiometric detection
  • Contains all necessary Components
  • Multiple applications
Caution: For research use only.