ContactUsLinkComponent
PhosphoWorks™ Colorimetric MESG Phosphate Assay Kit *UV Absorption*
PhosphoWorks™ Colorimetric MESG Phosphate Assay Kit *UV Absorption*
Catalog # AATB21659
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
PhosphoWorks™ Colorimetric MESG Phosphate Assay Kit *UV Absorption*
Catalog # AATB21659
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    PhosphoWorks™ colorimetric MESG phosphate assay kit *UV absorption*
Specifications
About this item
In the presence of inorganic phosphate MESG is converted to 2-amino-6-mercapto-7-methlpurine by purine nucleoside phosphorylase (EC 2.4.2.1) with absorption wavelength shift to red.
  • Provides all necessary components
  • Multiple applications
  • Photometric detection
Caution: For research use only.